Linearity Theraputic Drug Monitoring, AUDIT Microcontrols
Supplier: AUDIT MICROCONTROLS
K707M-5
97068-892PK
603.56
USD
97068-892
97068-916
Linearity Theraputic Drug Monitoring, AUDIT Microcontrols
Controls and Calibration Verification Materials
Universal TDM Linearity and Siemens Centaur TDM Linearity are assayed quality control materials consisting of five levels of human serum.
Each kit has five distinct levels that demonstrate a linear relationship to each other for their respective analytes, reagents and instruments.
Learn more
![](/m-us.vwr.com/en_US/images/about_vwr_275x315.jpg)