VWR® PCR-To-Gel Taq Master Mix, 2X
Supplier: VWR
1B1409-100RXN
97068-148EA 114.3 USD
97068-148 97068-150
VWR® PCR-To-Gel Taq Master Mix, 2X
Nucleic Acid Reagents End-point PCR Enzymes and Kits
Hot Start PCR-To-Gel Taq Master Mix, 2X is a ready-to-use reaction cocktail that contains all required components, except primers and template DNA, for routine PCR amplification of DNA fragments up to 4 kb
The included Taq DNA polymerase is inactivated with proprietary monoclonal antibodies. Incubation at ≥ 94°C during PCR thermal cycling irreversibly denatures the antibody and releases full Taq DNA polymerase activity. Inclusion of tracking dyes in the master mix allows direct loading and analysis of PCR products by agarose gel electrophoresis following PCR amplification.