Capto™ Butyl Hydrophobic Interaction Chromatography Media, Cytiva
Supplier: Cytiva
17-5459-01
97067-926EA
289.9
USD
97067-926
Capto™ Butyl Hydrophobic Interaction Chromatography Media, Cytiva
Protein Purification
Protein Purification Resins
Capto Butyl is a hydrophobic interaction chromatography (HIC) media used in the capture and intermediate stages of protein purification, Their combination of high capacity, high flow rate and low backpressure reduces process cycle times.
- Improved productivity and process economy in downstream operations
- Very high flow rates and large sample volume processing
- Excellent chemical stability