HiPrep Sephacryl S-500 HR Columns
Supplier: Cytiva
28-9356-06 28-9356-07
97067-772EA 1015.96 USD
97067-772 97067-774
HiPrep Sephacryl S-500 HR Columns
Chromatography Columns
HiPrep 26/60 Sephacryl S-500 HR are prepacked gel filtration column for preparative separation of biomolecules from up to 13 ml sample volume.
  • Gel filtration columns for low to medium pressure ÄKTA systems
  • Well-established in industrial production
  • The HiPrep Sephacryl series offers reproducible separations over a wide molecular weight range
  • Convenient, prepacked HiPrep Sephacryl columns speed up protein purification
Sephacryl S-500 is used for separating polysaccarides, macromolecules with extended structures, and even small plasmids. Maximum recovery and minimum nonspecific adsorption with long-term chemical and physical stability resulting from the hydrophilic, rigid allyl dextran/bisacrylamide matrix.