PlasmidSelect Xtra Hydrophobic Interaction Chromatography Media, Cytiva
Supplier: Cytiva
28-4024-02
97067-766EA 1315.17 USD
97067-766
PlasmidSelect Xtra Hydrophobic Interaction Chromatography Media, Cytiva
Protein Purification Protein Purification Resins
PlasmidSelect Xtra is a thiophilic aromatic adsorption chromatography medium with a selectivity that allows generic purification of supercoiled covalently closed circular forms of plasmid DNA to be separated from open circular forms.
  • Allows for generic purification of supercoiled plasmid DNA
  • Allows for consistent purification results when scaling up from research scale to cGMP production
  • Hydrophilic nature of the base matrix ensures low levels of non specific binding
  • BioProcess medium supported for industrial applications
Available in convenient prepacked format for research, process development and quantitative and qualitative analysis of supercoiled plasmid DNA.
Degree of substitution: Approx. 3.5 mg 2-mercaptopyridine/ml medium
Storage conditions: 4 to 30 °C, 20% Ethanol
Chemical stability: Stable in commonly used aqueous buffers