qPCR NGS Library Quantification Kit, Agilent Technologies
Supplier: AGILENT TECH
G4880A
97066-988KT 974.77 USD
97066-988
qPCR NGS Library Quantification Kit, Agilent Technologies
Assays Nucleic Acid Assays
The Agilent qPCR NGS Library quantification kit is a reliable method for quantifying NGS libraries.
  • Provides researchers with an accurate and sensitive method for quantifying NGS libraries
  • Validated in the SureSelect Target Enrichment Protocol for barcoding and indexing applications
  • Illumina and AB SOLID library quantification kits available
The Agilent qPCR NGS Library quantification kit provides researchers an accurate and sensitive method for quantifying NGS libraries. The NGS Library Quantification kit was validated in the SureSelect Target Enrichment Protocol for barcoding and indexing applications. Illumina and AB SOLID library quantification kits available.
Accurate library quantification leads to optimal cluster densities for improved sequence efficiency and data quality. Consistent quantification across a broad range of samples, varying library insert sizes, and GC content. Quantify 84 libraries with each kit.