Multiple Affinity Removal Spin Cartridge HSA/IgG, Agilent Technologies
Supplier: Agilent Technologies
5188-8825
97066-656EA 876.44 USD
97066-656
Multiple Affinity Removal Spin Cartridge HSA/IgG, Agilent Technologies
Chromatography Columns
The multiple affinity removal spin cartridge – Human Albumin/IgG is designed to remove two interfering high-abundant proteins from human samples (such as plasma, serum, urine or cerebrospinal fluid).
  • Compatible with standard benchtop microcentrifuges and luer-lok syringes
  • Serum load volume is 50 µl per run
  • Optimize sample recovery by utilizing our rapid, single injection protocols and optimized buffers
Removal of these abundant proteins improves the subsequent LC/MS and electrophoretic analysis of the sample by effectively expanding the dynamic range of the analysis.
Caution: For research use only. Not for use in diagnostic procedures.