Brilliant III SYBR® MM, Agilent Technologies
Supplier: AGILENT TECH
Brilliant III Ultra-Fast SYBR® QPCR and QRT-PCR Master Mixes deliver superior specificity by utilizing a novel faster-activating hot start method to minimize the formation of primer-dimers and other off-target reactions.
- Reliable and reproducible data
- Novel fast Taq mutant for qPCR results in under 35 minutes
- Enhanced rapid hot start capability saves time and reduces primer-dimer formation
- Convenient pre-blended formulations compatible with any SYBR Green dye
- Designed for researchers who want access to their data faster without compromising data quality
These unique reagents are equally robust and reproducible across a variety of assays on fast cycling real-time platforms.
Caution: For research use only. Not for use in diagnostic procedures.