Universal Reverse Primer, Agilent Technologies
Supplier: AGILENT TECH
The universal reverse primer serves as the downstream primer in the qPCR reaction by annealing to a universal sequence tag that is added to the cDNA during the reverse transcription protocol.
Applications: PCR
Number of Reactions: 3.125 μM
Physical Form: Liquid
Reactions per Kit: 300
Volume: 200 µL
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...