VWR® Vacuum Filtration Systems featuring Pall Membranes
Supplier: VWR International
1101-VWR 1102-VWR 1141-VWR 1142-VWR 1151-VWR 1152-VWR 1153-VWR 1161-VWR 1162-VWR 1163-VWR 1116-VWR 1122-VWR 1103-VWR 1143-VWR 116-7123-VWR 116-8123-VWR 116-9123-VWR 116-1123-VWR 116-2123-VWR 116-3123-VWR 146-5123-VWR 146-4123-VWR 1411-VWR 146-2323-VWR 146-1123-VWR 146-1223-VWR
97066-200CS 251.12 USD
97066-200 97066-202 97066-206 97066-208 97066-212 97066-214 97066-216 97066-218 97066-220 97066-222 97066-232 97066-234 97066-204 97066-210 89220-694 89220-696 89220-698 89220-700 89220-702 89220-704 89220-708 89220-714 89220-716 89220-724 89220-726 89220-728
VWR® Vacuum Filtration Systems featuring Pall Membranes
Filters Bottle Top Filters
VWR® Vacuum Filtration Systems are designed for the preparation of buffers, tissue culture media, microbiological media, and other biological fluids.
  • For the preparation of biological fluids
  • Feature Pall membranes
  • Compatible with the quick-connect vacuum pedestal for use with existing laboratory stands
They feature large diameter Pall membranes for guaranteed performance and maximum throughput.
The centrifuge tube filtration system includes a filter funnel, 15 or 50 ml centrifuge tube with polypropylene screw cap, and vacuum hose adapter. The system is designed to allow the user to filter fluids directly into the tube, eliminating the potential for contamination and spills during post-sterilization transfer. The bottle-top filtration system includes a receiver bottle with an ergonomic shape, molded finger grips, and a patented bottle cap.
All systems are compatible with the quick-connect vacuum pedestal, vacuum adapter (included), and cradle ring for use with existing laboratory stands. All three options allow for a convenient one-time vacuum hose connection, and one-handed operation. All items are available with 0.1 µm (Mycoplasma filtering grade), 0.2 µm (sterilizing grade) or 0.45 µm (clarification grade) asymmetric PES membranes. All items are made from Class VI materials, and are conveniently sold gamma sterilized.
Ordering information: Vacuum filtration pedestal and vacuum filtration cradle ring are sold separately.