Corning® Charcoal-Stripped FBS, Corning
Supplier: Corning
35-072-CV
97065-304EA 1453.32 USD
97065-304
Corning® Charcoal-Stripped FBS, Corning
Sera (Serum)
Charcoal-filtered (or “charcoal-stripped”) FBS is often used in immunoassays and hormone studies
Charcoal or charcoal-dextran removes lipophilic compounds (hormones, certain growth factors and cytokines) and virus particles.