Acetone ≥99.5% ACS

Supplier: VWR
Danger

Synonyms: 2-Propanone, DMK, Propanone, Dimethyl ketone

E646-1L E646-500ML E646-4L
97064-786CS 260.63 USD
97064-786 97065-030 97065-060
Acetone ≥99.5% ACS
Acetone
Formula: CH₃COCH₃
MW: 58.08 g/mol
Boiling Pt: 56.2
Melting Pt: -95.4
Density: 0.792 g/cm³ (20 °C)
Flash Pt: <–20 °C (closed cup)
Storage Temperature: Ambient
MDL Number: MFCD00008765
CAS Number: 67-64-1
EINECS: 200-662-2
UN: 1090
ADR: 3 II
REACH: 01-2119471330-49
Merck Index: 13,00067
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=3de5d8e3bf7a94692d684c8f0ccdfb8500fde76d&vid=24fb2e0ec86974b1803780c7d5b23804a085df64&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Abs.@330nm 1
Abs.@340nm = 0.1
Abs.@350nm = 0.02
Abs.@400nm = 0.01
Aldehyde = 0.002%
Color (APHA) = 10
Isopropyl Alcohol = 0.05%
Methanol = 0.05%
Purity = 99.5%
Residue After Evaporation = 0.001%
Solubility (50%, CO2 free Water) Pass
Substances Reducing Permanganate Test Pass
Titrable Acid = 0.0003 meq/g
Titrable Base = 0.0006 meq/g


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR