Potassium permanganate ACS

Supplier: VWR
Danger

0277-2.5KG
97065-026EA 255.77 USD
97065-026
Potassium permanganate ACS
Potassium permanganate
Formula: KMnO₄
Melting Pt: > 240 °C
Density: 2.7 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011364
CAS Number: 7722-64-7
EINECS: 231-760-3
UN: 1490
ADR: 5.1,II
Merck Index: 12,07824

Order Now

Specification Test Results

Chloride & Chlorate 0.005%
Insolubles = 0.2%
Purity = 99%
Sulfate = 0.02%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR