Acetonitrile ≥99.5% for HPLC
Supplier: VWR
|
Danger
|
Synonyms:
Methanecarbonitrile, Methane, cyano-, Methyl cyanide, Ethyl nitrile, ACN, Cyanomethane, Ethanenitrile, Ethanoic acid nitrile
K981-1L
K981-4L
97065-022CS
2025.48
USD
97065-022
97066-096
Acetonitrile ≥99.5% for HPLC
Acetonitrile
- High purity, functionally tested reagents suitable for HPLC and mass spectrometry.
- Low UV absorbing and low particulate count to reduce background.
Formula:
H₃CCN MW: 41.05 g/mol Boiling Pt: 81.6 °C (1013 hPa) Melting Pt: –45.7 °C Density: 0.782 g/cm³ (20 °C) Flash Pt: 2 °C (closed cup) Storage Temperature: Ambient |
MDL Number:
MFCD00001878 CAS Number: 75-05-8 EINECS: 200-835-2 UN: 1648 ADR: 3,II REACH: 01-2119471307-38 Merck Index: 13,00071 |
|
Specification Test Results
Abs.@220nm |
|
Abs.@254nm |
|
Assay |
|
Liquid Chromatography Suitability | Pass |
Residue After Evaporation |
|
Titrable Acid |
|
Titrable Base |
|
Water (KF) |
|
Learn more
![](/m-us.vwr.com/en_US/images/about_vwr_275x315.jpg)
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...