Bradford's reagent, Biotechnology Grade for quantitative protein determination
Supplier: VWR
|
Warning
|
E530-1L
97065-020CS
2248.38
USD
97065-020
Bradford's reagent, Biotechnology Grade for quantitative protein determination
Bradford's reagent
Specification Test Results
Abs.@ Peak A | 0.95 - 1.6 |
Abs.@ Peak A/Abs.@ 595nm |
|
Bradford Protein Assay | Pass |
Characteristic Pattern | Pass |
Peak A | 463 nm - 467 nm |
pH @25°C | 0.5 - 1.5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...