Ethidium bromide stock solution 10 mg/mL in aqueous solution, Biotechnology Grade

Supplier: VWR
Warning

X328-10ML
97064-970EA 70.1 USD
97064-970
Ethidium bromide stock solution 10 mg/mL in aqueous solution, Biotechnology Grade
Ethidium bromide

Ethidium Bromide solution is a fluorescent stain for visualizing nucleic acids in solution or in electrophoresis gels. At a concentration of 0.5 µg/ml, ethidium bromide is useful in agarose or acrylamide ges for band detection. It is used to detect as little as 10 ng of nucleic acid. At high concentrations (0.5 - 1.0 µg/ml), it may be used to facilitate purification of DNA in cesium chloride gradient.

Formula: C₂₁H₂₀BrN₃
MDL Number: MFCD00011724
CAS Number: 1239-45-8
EINECS: 214-984-6

Order Now

Specification Test Results

DNase none detected
Moisture = 8%
Purity (Dry Basis) = 98%
RNase none detected


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR