Imidazole ≥99%, Proteomics Grade

Supplier: VWR
Danger

Synonyms: Glyoxaline, 1H-Imidazole, Glyoxalamine

M136-100G
97064-890CS 3372.39 USD
97064-890
Imidazole ≥99%, Proteomics Grade
Imidazole
Formula: C₃H₄N₂
MW: 68.08 g/mol
Boiling Pt: 255
Melting Pt: 89…90 °C
Density: 1.18 g/cm³ (20 °C)
Flash Pt: 135
Storage Temperature: Ambient
MDL Number: MFCD00005183
CAS Number: 288-32-4
EINECS: 206-019-2
UN: 3263
ADR: 8 II
Merck Index: 13,04935
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=48eba49e61201d6fffa2bef0c54633a20660c7ef&vid=e7f154e4976b52732539e49bbff60454d5eb82ff&prid=web&clid=SAPDAM

Order Now

Specification Test Results

DNase none detected
Appearance White to slightly yellow solid
Insolubles = 0.1 %
Melting Point 88°C - 92°C
pH (5%, Water) @25°C 9.5 - 10.5
Protease none detected
Purity = 99 %
RNase none detected
Solubility (20%, Water) Pass


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR