Acrylamide 40% in aqueous solution, Acryl 40™

Supplier: VWR
Danger

M121-500ML
97064-870EA 148.67 USD
97064-870
Acrylamide 40% in aqueous solution, Acryl 40™
Acrylamide (monomer)

VWR offers an extensive line of acrylamide and bis-acrylamide pre-weighed powder blends, pre-mixed stock solutions, and ready-to-use solutions for customized polyacrylamide gel electrophoresis of proteins and PAGE of nucleic acids. Our ultra pure (> 99.9%) acrylamide and bis-acrylamide powders provide the flexibility to prepare solutions having concentrations and ratios for all electrophoresis applications. Liquid blends minimize the handling of neurotoxic acrylamide.


  • Ultra-pure powders with acrylamide purity > 99.9%
  • Convenient ready-to-use
  • Eliminates hazardous powdered acrylamide handling

Formula: C₃H₅NO
MDL Number: MFCD00008032
CAS Number: 79-06-1
UN: 3426
ADR: 6.1,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=7687b7f17a8cdf31c559a33fd5d4021a2e50a40a&vid=fecebe0c34b037788df119c11bfc9637d78cda0d&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Acrylic Acid 0.005%
Conductivity 5 umhos
DNase none detected
Electrophoresis Pass
pH @25°C 6 - 7
Protease none detected
RNase none detected


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR