Acrylamide, Proteomics grade for biochemistry

Supplier: VWR
Danger

Synonyms: Prop-2-enamide

M120-500G
97064-866CS 3466.49 USD
97064-866
Acrylamide, Proteomics grade for biochemistry
Acrylamide (monomer)

White free-flowing, uniform crystals.


  • Ultra-pure powders with acrylamide purity > 99.9%
  • Convenient ready-to-use
  • Eliminates hazardous powdered acrylamide handling

Formula: C₃H₅NO
MW: 71.08 g/mol
Boiling Pt: 232
Melting Pt: 84.5
Density: 1.122 g/cm³ (20 °C)
Flash Pt: 138
Storage Temperature: Refrigerator
MDL Number: MFCD00008032
CAS Number: 79-06-1
EINECS: 201-173-7
UN: 3426
ADR: 6.1 III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=7687b7f17a8cdf31c559a33fd5d4021a2e50a40a&vid=fecebe0c34b037788df119c11bfc9637d78cda0d&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Conductivity (40%, Water) 5 umhos
DNase none detected
Electrophoresis Pass
Free Acrylic Acid 0.001 %
Appearance White free-flowing, uniform crystals
Insolubles 0.005 %
Iron 0.0001 %
Lead 0.0001 %
Magnesium 0.0001 %
Melting Point 84°C - 86°C
Moisture (KF) 0.2 %
pH (10%, 0.1M NaCl) @25°C 6.5 %
Protease none detected
Purity = 99.9%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR