Sodium dodecyl sulfate (SDS), Proteomics Grade

Supplier: VWR
Warning

M112-500ML M111-100ML M107-250G M107-100G
97063-772EA 132.49 USD
97063-772 97063-768 97064-860 97064-858 97065-004
Sodium dodecyl sulfate (SDS), Proteomics Grade
Sodium dodecyl sulphate (SDS)

Negatively charged anionic detergents are strong detergents utilized in lysis buffers for the complete disruption of membranes and denaturation of proteins. Anionic detergents are also used in denaturing gel electrophoresis.


  • High purity and protease-free
  • Suitable for most biological applications

Formula: CH₃(CH₂)₁₁OSO₃Na
MDL Number: MFCD00036175
CAS Number: 151-21-3
UN: 1325
ADR: 4.1,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=fe4cb1741fba41f699a3be006d0b01b1b551898c&vid=c6df9a86fc351b319e68cc79c1c17f733b4fec67&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR