Phenol : chloroform : iso-amyl alcohol (25:24:1) (v/v/v) ≥99.0%, Biotechnology Grade 1 Phase, VWR®
Supplier: VWR
|
Danger
|
Synonyms:
Carbolic acid, Hydroxybenzene, Phenic acid
K169-400ML
K169-100ML
97064-826CS
1574.98
USD
97064-826
97064-824
Phenol : chloroform : iso-amyl alcohol (25:24:1) (v/v/v) ≥99.0%, Biotechnology Grade 1 Phase, VWR®
Phenol : Chloroform : iso-Amyl alcohol
Storage Temperature:
Ambient |
MDL Number:
MFCD00133763 CAS Number: 136112-00-0 UN: 2810 ADR: 6.1 II |
Specification Test Results
Conductivity(50%, Water)@25°C | 250 uS - 330 uS |
pH(50%, Water)@25°C | 8.05 - 8.35 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...