NaOH/SDS Lysis solution, Ultra Pure Grade

Supplier: VWR

J611-500ML
97064-814EA 180.74 USD
97064-814
NaOH/SDS Lysis solution, Ultra Pure Grade
NaOH/SDS Lysis

An ultra pure reagent for alkaline lysis of bacterial cells used in standard DNA purification procedures.

Storage Temperature: Ambient
UN: 1824
ADR: 8,III

Order Now

Specification Test Results

Conductivity (10%, Water) @25C Passes Test
Titration 7.6 - 8.4 g/L


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR