Acetone ≥99.5% ACS
Supplier: VWR
|
Danger
|
Synonyms:
2-Propanone, DMK, Propanone, Dimethyl ketone
E646-1L
E646-500ML
E646-4L
97064-786CS
260.63
USD
97064-786
97065-030
97065-060
Acetone ≥99.5% ACS
Acetone
Formula:
CH₃COCH₃ MW: 58.08 g/mol Boiling Pt: 56.2 °C (1013 hPa) Melting Pt: –95.4 °C Density: 0.792 g/cm³ (20 °C) Flash Pt: <–20 °C (closed cup) Storage Temperature: Ambient |
MDL Number:
MFCD00008765 CAS Number: 67-64-1 EINECS: 200-662-2 UN: 1090 ADR: 3,II REACH: 01-2119471330-49 Merck Index: 13,00067 |
|
Specification Test Results
Abs.@330nm |
|
Abs.@340nm |
|
Abs.@350nm |
|
Abs.@400nm |
|
Aldehyde |
|
Color (APHA) |
|
Isopropyl Alcohol |
|
Methanol |
|
Purity |
|
Residue After Evaporation |
|
Solubility (50%, CO2 free Water) | Pass |
Substances Reducing Permanganate Test | Pass |
Titrable Acid |
|
Titrable Base |
|
Learn more
![](/m-us.vwr.com/en_US/images/about_vwr_275x315.jpg)
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...