Sodium hydroxide 10 N in aqueous solution, Reagent Grade, VWR®

Supplier: VWR

Synonyms: Caustic soda, Caustic soda, Sodium hydrate, Lye, Soda lye

E584-500ML E584-100ML
97064-784CS 473.32 USD
97064-784 97064-782
Sodium hydroxide 10 N in aqueous solution, Reagent Grade, VWR®
Sodium hydroxide
Formula: NaOH
MW: 40 g/mol
Boiling Pt: 109 °C (1013 hPa)
Density: 1.34 g/cm³ (20 °C)
MDL Number: MFCD00003548
CAS Number: 1310-73-2
EINECS: 215-185-5
UN: 1824
ADR: 8,II

Order Now

Specification Test Results

Acid Titration 9900 mmol/L - 10100 mmol/L

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR