Ethanol denatured, anhydrous 94.9-96.0% for biotechnology

Supplier: VWR

E193-500ML
97064-736CS 710.53 USD
97064-736 97065-056
Ethanol denatured, anhydrous 94.9-96.0% for biotechnology
Ethanol denatured

Denatured with 5% methanol

Formula: H₃CCH₂OH
Storage Temperature: Ambient
MDL Number: MFCD00003568
CAS Number: 64-17-5
UN: 1170
ADR: 3,II

Order Now

Specification Test Results

Identification PASS
Methanol 5.0 %
Moisture 1.0 %
Purity 94.9 - 96.0 %
Specific Gravity 0.7964


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR