Chloroform stabilized for biotechnology

Supplier: VWR
Danger

0757-500ML
97064-680CS 425.99 USD
97064-680 97064-678
Chloroform stabilized for biotechnology
Chloroform
Formula: CHCl₃
Boiling Pt: 59.4…61.5 °C
Melting Pt: –63 °C
Density: 1.4832 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00000826
CAS Number: 67-66-3
EINECS: 200-663-8
UN: 1888
ADR: 6.1,III
REACH: 05-2114614906-45
Merck Index: 13,02160
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=9cc00da8cefd4e04c0ea1d7c5801308cd73eeef9&vid=c7d46a2ee73651eae0f3654c272e820106b2f04f&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Abs.@ 245nm = 1
Abs.@ 255nm = 0.25
Abs.@ 260nm = 0.15
Abs.@ 270nm = 0.05
Abs.@ 290nm = 0.01
Abs.@ 400nm 0.01
Chloride 10 ppm
Color (APHA) 10
Lead 0.05 ppm
Moisture (KF) 0.05 %
Purity 99.8 %
Residue After Evaporation 0.001 %
Titratable Free Acid 0.5 meq/g


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR