Sodium azide, high purity
Supplier: VWR
|
Danger
|
Synonyms:
Sodium triazide
0639-250G
97064-646CS
1141.54
USD
97064-646
Sodium azide, high purity
Sodium azide
Formula:
NaN₃ MW: 65.01 g/mol Melting Pt: 275 °C Density: 1.85 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00003536 CAS Number: 26628-22-8 EINECS: 247-852-1 UN: 1687 ADR: 6.1,II Merck Index: 13,08653 |
Learn more
![](/m-us.vwr.com/en_US/images/about_vwr_275x315.jpg)
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...