1,10-Phenanthroline monohydrate ACS

Supplier: VWR
Danger

0516-10G
97064-614CS 827.72 USD
97064-614
1,10-Phenanthroline monohydrate ACS
1,10-Phenanthroline monohydrate

A metalloprotease inhibitor useful for chelating iron and other divalent metals.

Formula: C₁₂H₈N₂·1H₂O
Melting Pt: 93…94 °C
Density: 1.259 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00149973
CAS Number: 5144-89-8
EINECS: 200-629-2
UN: 2811
ADR: 6.1,III
Merck Index: 13,07291
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=aba346b9f67fdda9afd4d48b893e6318564d604c&vid=2d08e04ee9525293a747b55cdcff8f66a6c4715a&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Suitability as Redox Indicator Pass
Suitability for Determining Iron pass


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR