Sodium selenite, high purity

Supplier: VWR
Danger

0321-100G
97064-558CS 1023.9 USD
97064-558
Sodium selenite, high purity
Sodium selenite
Formula: Na₂SeO₃
MW: 172.94 g/mol
Melting Pt: 710
Density: 3.1 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00003489
CAS Number: 10102-18-8
EINECS: 233-267-9
UN: 2630
ADR: 6.1 I
Merck Index: 12,08822

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR