Sodium hydroxide, beads, Reagent Grade

Supplier: VWR
Danger

Synonyms: Caustic soda, Caustic soda, Sodium hydrate, Lye, Soda lye

0173-1KG 0173-5KG
97064-526CS 648.72 USD
97064-526 97064-528
Sodium hydroxide, beads, Reagent Grade
Sodium hydroxide
Formula: NaOH
MW: 40 g/mol
Boiling Pt: 1390 °C (1013 hPa)
Melting Pt: 323 °C
Density: 2.13 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00003548
CAS Number: 1310-73-2
EINECS: 215-185-5
UN: 1823
ADR: 8,II
REACH: 01-2119457892-27
Merck Index: 14,08627

Order Now

Specification Test Results

Chloride <= 0.05 %
Heavy Metals <= 0.006 %
Potassium <= 0.1 %
Purity >98.5 %
Sodium Carbonate <= 0.90 %
Sulfate <= 0.005 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR