Sodium hydroxide, pellets ACS

Supplier: VWR
Danger

Synonyms: Caustic soda, Caustic soda, Sodium hydrate, Lye, Soda lye

0583-500G 0583-50KG 0583-5KG 0583-1KG 0583-12KG
97064-476CS 688.94 USD
97064-476 97064-478 97064-480 97064-486 97064-498
Sodium hydroxide, pellets ACS
Sodium hydroxide
Formula: NaOH
Boiling Pt: 1390 °C (1013 hPa)
Melting Pt: 323 °C
Density: 2.13 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00003548
CAS Number: 1310-73-2
EINECS: 215-185-5
UN: 1823
ADR: 8,II
REACH: 01-2119457892-27
Merck Index: 14,08627

Order Now

Specification Test Results

Calcium 0.005%
Chloride = 0.005%
Heavy Metals = 0.002%
Iron = 0.001%
Magnesium = 0.002%
Mercury = 0.1 ppm
Nickel = 0.001%
Nitrogen Compounds = 0.001%
Phosphate = 0.001%
Potassium = 0.02%
Purity = 97%
Sodium Carbonate = 1%
Sulfate = 0.003%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR