Sodium dodecyl sulfate (SDS) ≥99%, Biotechnology Grade for biotechnology
Supplier: VWR
|
Danger
|
Synonyms:
Sodium lauryl sulfate, Dodecyl sulfate sodium salt, Lauryl sulfate sodium salt, Dodecyl sulfuric acid sodium salt, Dodecyl sulphate sodium salt, Sodium dodecyl sulfate, Dodecyl sulphuric acid sodium salt, Lauryl sulphate sodium salt, Sodium lauryl sulphate
0227-250G
0227-500G
0227-12KG
0227-1KG
0227-5KG
0227-100G
97064-472CS
1750.73
USD
97064-472
97064-474
97064-504
97064-470
97064-494
97064-496
Sodium dodecyl sulfate (SDS) ≥99%, Biotechnology Grade for biotechnology
Sodium dodecyl sulphate (SDS)
For applications ranging from solubilizing membrane proteins in non-denaturing conditions to creating denaturing conditions during gel electrophoresis.
Formula:
CH₃(CH₂)₁₁OSO₃Na Melting Pt: 204 …207 °C Density: 1.1 g/cm³ (20 °C) Flash Pt: >100 °C Storage Temperature: Ambient |
MDL Number:
MFCD00036175 CAS Number: 151-21-3 EINECS: 205-788-1 UN: 1325 ADR: 4.1,III Merck Index: 14,08636 |
|
Specification Test Results
Abs.@260nm (3%, Water) |
|
Abs.@280nm (3%, Water) |
|
Chloride |
|
Copper |
|
DNase | none detected |
Heavy Metals (as Pb) |
|
Iron |
|
Phosphate |
|
Protease | none detected |
Purity |
|
RNase | none detected |
Solubility (10%, Water) | Pass |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...