Sodium dodecyl sulfate (SDS) ≥99%, Biotechnology Grade for biotechnology

Supplier: VWR
Danger

Synonyms: Sodium lauryl sulfate, Dodecyl sulfate sodium salt, Lauryl sulfate sodium salt, Dodecyl sulfuric acid sodium salt, Dodecyl sulphate sodium salt, Sodium dodecyl sulfate, Dodecyl sulphuric acid sodium salt, Lauryl sulphate sodium salt, Sodium lauryl sulphate

0227-250G 0227-500G 0227-12KG 0227-1KG 0227-5KG 0227-100G
97064-472CS 1750.73 USD
97064-472 97064-474 97064-504 97064-470 97064-494 97064-496
Sodium dodecyl sulfate (SDS) ≥99%, Biotechnology Grade for biotechnology
Sodium dodecyl sulphate (SDS)

For applications ranging from solubilizing membrane proteins in non-denaturing conditions to creating denaturing conditions during gel electrophoresis.

Formula: CH₃(CH₂)₁₁OSO₃Na
Melting Pt: 204 …207 °C
Density: 1.1 g/cm³ (20 °C)
Flash Pt: >100 °C
Storage Temperature: Ambient
MDL Number: MFCD00036175
CAS Number: 151-21-3
EINECS: 205-788-1
UN: 1325
ADR: 4.1,III
Merck Index: 14,08636
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=fe4cb1741fba41f699a3be006d0b01b1b551898c&vid=c6df9a86fc351b319e68cc79c1c17f733b4fec67&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Abs.@260nm (3%, Water) 0.1
Abs.@280nm (3%, Water) = 0.1
Chloride = 0.1%
Copper = 0.0005%
DNase none detected
Heavy Metals (as Pb) = 0.0005%
Iron = 0.0001%
Phosphate = 0.0005%
Protease none detected
Purity = 99%
RNase none detected
Solubility (10%, Water) Pass


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR