3,3',5,5'-Tetramethylbenzidine (TMB), liquid, Biotechnology Grade 1 component

Supplier: VWR
Danger

Synonyms: TMB, 3,3',5,5'-Tetramethyl-[1,1'-biphenyl]-4,4'-diamine

J644-100ML
97064-344CS 1150.19 USD
97064-344
3,3',5,5'-Tetramethylbenzidine (TMB), liquid, Biotechnology Grade 1 component
3,3',5,5'-Tetramethylbenzidine (TMB)

Ready-to-use sensitive substrate for the detection of horseradish peroxidase activity


Absorbs at 450 nm (yellow end-product). Ideal for ELISA and solution assays.

Formula: C₁₆H₂₀N₂
MW: 240.35 g/mol
MDL Number: MFCD00007748
CAS Number: 54827-17-7

Order Now

Specification Test Results

Abs.@650nm < 0.02
pH @25°C 5 - 7
UV/Vis Assay Pass

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR