Puromycin dihydrochloride, Ultra Pure Grade

Supplier: VWR
Warning

Synonyms: (S)-2-Amino-N-((2S,3S,4R,5R)-5-(6-(Dimethylamino)-9H-purin-9-yl)-4-hydroxy-2-(hydroxymethyl)tetrahydrofuran-3-yl)-3-(4-methoxyphenyl)propanamide dihydrochloride

J593-25MG
97064-280EA 68.45 USD
97064-280
Puromycin dihydrochloride, Ultra Pure Grade
Puromycin dihydrochloride

A broad spectrum antibiotic that inhibits protein synthesis in both procaryotic and eucaryotic organisms


Widely used as a selection agent for cells carrying the Pac resistance gene encoding puromycin N-acetyl-transferase.

Formula: C₂₂H₂₉N₇O₅·2HCl
MW: 544.44 g/mol
MDL Number: MFCD00012691
CAS Number: 58-58-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR