D-(+)-Mannose, high purity

Supplier: VWR

Synonyms: (2S,3S,4R,5R)-2,3,4,5,6-Pentahydroxyhexanal

J443-500G
97064-178CS 1405.85 USD
97064-178 97064-182
D-(+)-Mannose, high purity
D(+)-Mannose
Formula: C₆H₁₂O₆
Boiling Pt: 376 °C (1013 hPa)
Melting Pt: 132 °C (dec.)
Density: 1.539 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00064122
CAS Number: 3458-28-4
EINECS: 222-392-4
Merck Index: 13,05772

Order Now

Specification Test Results

Loss on Drying 1%
Melting Point 125°C - 131°C
Purity (HPLC) 99.5 %
Solubility (5%, Water) Pass
Specific Rotation 14° - 15°


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR