VWR® TAQ DNA Polymerase
Supplier: VWR
N224-500U
97064-160EA 526.33 USD
97064-160 97064-234
VWR® TAQ DNA Polymerase
Nucleic Acid Reagents End-point PCR Enzymes and Kits
Taq DNA Polymerase is ideal for general PCR and primer extension reactions to amplify DNA templates with high sensitivity.
  • Amplify DNA templates with high sensitivity
  • Amplify single copy & low abundance templates
  • Nuclease-free, high-purity reagents
The PCR Kit with Taq DNA Polymerase includes separate solutions of all material needed for a PCR reaction, including dNTPs, reaction buffer with and without MgCl2, 25mM MgCl2, Taq Polymerase, and sterile water. Taq DNA Polymerase is ideal for general PCR and primer extension reactions to amplify DNA templates with high sensitivity. This DNA polymerase is highly-purified, DNA-free, and free of bacterial genomic DNA. PCR can be initiated from as few as 6 primed templates. One unit of activity incorporates 10 nmoles of dNTP into acid-insoluble DNA in 30 minutes at 72ºC. It contains sufficient polymerase for 50 amplifications.
Taq DNA Polymerase: 500 U + DFS Taq Buffer, 10X, 1ml
PCR kit with Taq DNA Polymerase includes:
10 mM dNTPs, 50X 10X Reaction Buffer with MgCl2
10X Reaction Buffer without MgCl2
25 mM MgCl2
Taq DNA Polymerase, 500 U
Sterile, ultrapure water, 2x1.5 ml