VWR® Phosphate Buffered Saline, Ultra Pure Grade
Supplier: VWR
J373-4L
97064-158EA
219.54
USD
97064-158
VWR® Phosphate Buffered Saline, Ultra Pure Grade
Buffers
Cell Culture Buffers
1X Solution Composition: 137 mM NaCl, 2.7 mM KCl, 9.8 mM Phosphate buffer
- Preparation for use as 1X PBS, Dilute 1:10 in deionized water.
- May be sterilized by filtration or autoclave, if desired.
- DNase, RNase,Protease none detected.
- Conductivity at 25°C (1X PBS):14,000-17800 umhos
- pH at 25°(1X PBS): 7.3 – 7.5
Does not contain magnesium or calcium.
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...