Magnesium chloride, anhydrous, high purity

Supplier: VWR

J364-500G J364-100G J364-1KG
97063-152CS 1340 USD
97063-152 97064-150 97064-152
Magnesium chloride, anhydrous, high purity
Magnesium chloride
Formula: MgCl₂
Boiling Pt: 1412 °C (1013 hPa)
Melting Pt: 715 °C
Density: 2.32 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011106
CAS Number: 7786-30-3
EINECS: 232-094-6
Merck Index: 13,05684

Order Now

Specification Test Results

Insolubles 0.2%
Moisture (KF) 2 %
Purity 98 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR