TRITON™ X-100 (Polyethylene glycol tert-octylphenyl ether) 10% in aqueous solution, Proteomics Grade peroxide-free

Supplier: VWR
Danger

Synonyms: Polyethyleneglycol-[4-(1,1,3,3-tetramethylbutyl)phenyl]-ether, (p-tert-Octylphenoxy)polyethoxyethanol, Octoxynol, Polyoxyethylene(10) Octylphenyl Ether, t-Octylphenoxypolyethoxyethanol, O-[4-(1,1,3,3-Tetramethylbutyl)phenoxy]polyethoxyethanol

M236-10ML-5PK
97063-996EA 171.9 USD
97063-996
TRITON™ X-100 (Polyethylene glycol tert-octylphenyl ether) 10% in aqueous solution, Proteomics Grade peroxide-free
TRITON™ X-100 (Polyethylene glycol tert-octylphenyl ether)

Detergent


  • Optimized for isolating proteins and sample preparation
  • Contains < 1ug of peroxide per ml
  • Convenient concentrations pre-measured in ampoules
  • Long-term stability


VWR’s high purity Pro-PureTM grade detergents are carefully packed under inert gas, making them extremely stable. The detergents are provided in ready-to-use 10% solutions that are packaged in 10 ml ampoules for optimal convenience.

MDL Number: MFCD00132505
CAS Number: 9002-93-1

Order Now

Specification Test Results

Color (APHA) ≤100
Peroxides-free Pass/fail
pH (50%, Water, 25° C) 6.0 - 8.0
Protease- free Pass/fail

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR