Tween® 20 (Polysorbate) 10% in aqueous solution, Proteomics Grade peroxide-free

Supplier: VWR

M228-10ML-5PK
97063-980EA 172.21 USD
97063-980
Tween® 20 (Polysorbate) 10% in aqueous solution, Proteomics Grade peroxide-free
Tween® 20 (Polysorbate)

  • Optimized for isolating proteins and sample preparation
  • Contains < 1ug of peroxide per ml
  • Convenient concentrations pre-measured in ampoules
  • Long-term stability


VWR’s high purity Pro-PureTM grade detergents are carefully packed under inert gas, making them extremely stable. The detergents are provided in ready-to-use 10% solutions that are packaged in 10 ml ampoules for optimal convenience.

Formula: C₅₈H₁₁₄O₂₆
Storage Temperature: Ambient
MDL Number: MFCD00165986
CAS Number: 9005-64-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR