Milk, non fat (skimmed milk), powder, Proteomics Grade

Supplier: VWR

M203-10G-10PK
97063-958EA 103.45 USD
97063-958
Milk, non fat (skimmed milk), powder, Proteomics Grade
Non-fat milk powder (skim milk)

10 g per pack

Storage Temperature: Ambient
CAS Number: 999999-99-4
EINECS: 310-127-6

Order Now

Specification Test Results

Moisture (LOD) 4%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR