VWR® SDS-Page Gel Buffers
Supplier: VWR
M197-500ML
97063-954EA
224.49
USD
97063-954
97063-956
VWR® SDS-Page Gel Buffers
Buffers
Electrophoresis Buffers
Running Buffers
The 4X concentrated buffer (0.5M Tris pH 6.8 and 0.4% SDS) for preparation of stacking gels for Laemmli SDS-Page
- Reduce pipetting steps with convenient 4X solutions
Dilution to a 1X solution results in final 1X concentrations of 0.125M Tris and 0.1% SDS .