Tris(hydroxymethyl)aminomethane (TRIS, Trometamol), Proteomics Grade
Supplier: VWR
|
Warning
|
Synonyms:
2-Amino-2-(hydroxymethyl)propane-1,3-diol(tris), Tromethamine, Tris(hydroxymethyl)methylamine, 2-Amino-2-(hydroxymethyl)propane-1,3-diol, 2-Amino-2-(hydroxymethyl)-1,3-propanediol, Trizma(R) base, TRIS base, THAM
M151-1KG
M151-500G
97063-888CS
630.81
USD
97063-888
97063-890
Tris(hydroxymethyl)aminomethane (TRIS, Trometamol), Proteomics Grade
Tris(hydroxymethyl)aminomethane (TRIS, Trometamol)
Formula:
NH₂C(CH₂OH)₃ MW: 121.14 g/mol Boiling Pt: 288 °C Melting Pt: 169 °C Density: 1.328 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00004679 CAS Number: 77-86-1 EINECS: 201-064-4 Merck Index: 13,09842 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...