VWR® Trypsin, Crystalline, Proteomics Grade

Supplier: VWR

M150-1G
97063-884EA 96.86 USD
97063-884
VWR® Trypsin, Crystalline, Proteomics Grade
Enzymes

Trypsin, Crystalline, Proteomics Grade.


  • EINECS: 232-650-8
  • GHS Symbol 1: GHS08
  • GHS Symbol 2: GHS07
  • MDL: MFCD00082094

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR