Sodium thiosulfate pentahydrate, Proteomics Grade

Supplier: VWR

M132-500G
97063-834EA 34.52 USD
97063-834
Sodium thiosulfate pentahydrate, Proteomics Grade
Sodium thiosulphate pentahydrate
Formula: Na₂S₂O₃.5H₂O
Boiling Pt: 100 °C
Melting Pt: 49 °C
Density: 1.67 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00149186
CAS Number: 10102-17-7
EINECS: 231-867-5
UN: 0
ADR: ,
Merck Index: 12,08844

Order Now

Specification Test Results

Insolubles 0.005%
pH (5%, Water) @25°C 6 - 8.4
Protease none detected
Purity 99 %
Sulfate & Sulfite 0.1 %
Sulfide 0.0001 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR