Urea 8 M in aqueous solution, Proteomics Grade

Supplier: VWR

Synonyms: Carbonyl diamide, Carbamide, Diaminomethanone, Carbonyldiamide

M124-250ML
97063-804EA 153.29 USD
97063-804
Urea 8 M in aqueous solution, Proteomics Grade
Urea

A convenient pre-mixed solution that is easily diluted for buffer preparation under denaturing conditions.Commonly used to denature nucleic acids for electrophoresis and to study the secondary and tertiary structure of proteins. Effective for maintaining DNA in a highly denatured state during sequencing protocols, urea is also useful for accelerating the activity of Proteinase K digestions.

Formula: NH₂CONH₂
MW: 60.06 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00008022
CAS Number: 57-13-6
EINECS: 200-315-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=a9682f71b212a449398841e2beace2d7500b4710&vid=d6057a7ec3d606031d338a300b5d5db30b52d861&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Clarity Pass
Protease none detected
Urea 7.8 Molar - 8.2 Molar
Appearance Clear and colorless liquid

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR