Urea, Proteomics Grade

Supplier: VWR

Synonyms: Carbonyl diamide, Carbamide, Diaminomethanone, Carbonyldiamide

M123-1KG M123-500G
97063-798CS 478.76 USD
97063-798 97063-802
Urea, Proteomics Grade
Urea

Commonly used to denature nucleic acids for electrophoresis and to study the secondary and tertiary structure of proteins. Effective for maintaining DNA in a highly denatured state during sequencing protocols, urea is also useful for accelerating the acti

Formula: NH₂CONH₂
MW: 60.06 g/mol
Boiling Pt: 197
Melting Pt: 132.7
Density: 1.34 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00008022
CAS Number: 57-13-6
EINECS: 200-315-5
REACH: 01-2119463277-33
Merck Index: 13,09935
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=a9682f71b212a449398841e2beace2d7500b4710&vid=d6057a7ec3d606031d338a300b5d5db30b52d861&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Abs.@280nm (5M, Water) 0.05
Biuret Test Pass
Chloride = 0.0005 %
Cyanate = 0.002 %
DNase none detected
Heavy Metals (as Pb) = 0.0005 %
Insolubles 0.005 %
Iron = 0.0001 %
Melting Point 132°C - 135°C
Protease none
Purity = 99.5 %
RNase none detected
Solubility (5M, Water) Pass


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR