TRIS HCl (tris(hydroxymethyl)aminomethane hydrochloride), Proteomics Grade

Supplier: VWR
Warning

M108-1KG
97063-756CS 1371.14 USD
97063-756 97063-754
TRIS HCl (tris(hydroxymethyl)aminomethane hydrochloride), Proteomics Grade
TRIS HCl (Tris-(hydroxymethyl) aminomethane hydrochloride)
Formula: NH₂C(CH₂OH)₃·HCl
Melting Pt: 150…152 °C
Storage Temperature: Ambient
MDL Number: MFCD00012590
CAS Number: 1185-53-1
EINECS: 214-684-5
Merck Index: 12,09902
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=3cbdc9446fdb9bcfe3f1d992ff20a16726f98113&vid=3d668801395b81fa595401a752279006da8fd3cb&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Calcium 0.0001%
Copper 0.0001 %
DNase none detected
Electrophoresis Pass
Insolubles 0.001 %
Iron 0.001 %
Lead 0.0001 %
Magnesium 0.0001 %
Manganese 0.0001 %
pH (0.1M, Water) @25°C 4.2 - 4.9
Protease none detected
Purity 99.5
RNase none detected
Zinc = 0.0001%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR