Glycine, Proteomics Grade

Supplier: VWR

Synonyms: Aminoacetic acid, Glycocoll, H-Gly-OH

M103-5KG M103-1KG
97063-738EA 409.06 USD
97063-738 97063-736
Glycine, Proteomics Grade
Glycine
Formula: NH₂CH₂COOH
Boiling Pt: 240 °C
Melting Pt: 83 °C
Density: 1.64 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00008131
CAS Number: 56-40-6
EINECS: 200-272-2
Merck Index: 13,04504
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=d944007d30d5d4650062014a1b152bd9bfb66ea3&vid=093f4227cbdd1c4c6515ef9c4b85d93799877dcc&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR